Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PPAR delta Antibodies

          Invitrogen

          PPAR delta Polyclonal Antibody

          Advanced Verification
          20 Published Figures
          36 References
          View all (20) PPAR delta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PPAR delta Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (20)
          • Advanced Verification (3)
          PPAR delta Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PPAR delta Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 27

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPAR delta Antibody (PA1-823A) in ICC/IF

          Immunofluorescence analysis of PPAR delta was done on 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with PPAR delta Rabbit Polyclonal Antibody (Product # PA1-823A) at 1 µg/mL in 1% BSA and incubated for 3 hours at room tempe... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPAR delta Antibody in Immunocytochemistry (ICC/IF)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PPAR delta Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PPAR delta Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PPAR delta Antibody in Immunohistochemistry (IHC)
          PPAR delta Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PPAR delta Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PPAR delta Antibody in Immunohistochemistry (IHC)
          PPAR delta Antibody in Immunohistochemistry (IHC)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot (WB)
          PPAR delta Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PPAR delta Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PPAR delta Antibody in ChIP Assay (ChIP)
          PPAR delta Antibody
          PPAR delta Antibody
          PPAR delta Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PPAR delta Polyclonal Antibody

          Product Details

          PA1-823A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:250-1:1,000
          View 24 publications 24 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:200
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues M(1) E Q P Q E E T P E A R E E(14) C of mouse PPAR delta.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2165895

          Product Specific Information

          PA1-823A detects peroxisome proliferator activated receptor (PPAR) delta from rat, human and mouse cells. This antibody does not detect PPAR alpha or PPAR gamma.

          PA1-823A has been successfully used in Western blot and immunohistochemical and immunofluoresence procedures. By Western blot, this antibody detects an ~49 kDa protein representing PPAR delta from NIH-3T3 cell lysate. Immunolocalization yeilds predominant nuclear staining.

          The PA1-823A immunogen is a synthetic peptide corresponding to residues M(1) E Q P Q E E T P E A R E E(14) C of mouse PPAR delta. This sequence is 86% conserved in human PPAR delta. PA1-823A immunizing peptide (Cat. # PEP-026) is available for use in neutralization and control experiments.

          Target Information

          This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) family. PPARs are nuclear hormone receptors that bind peroxisome proliferators and control the size and number of peroxisomes produced by cells. PPARs mediate a variety of biological processes, and may be involved in the development of several chronic diseases, including diabetes, obesity, atherosclerosis, and cancer. This protein is a potent inhibitor of ligand-induced transcription activity of PPAR alpha and PPAR gamma. It may function as an integrator of transcription repression and nuclear receptor signaling. The expression of this gene is found to be elevated in colorectal cancer cells. The elevated expression can be repressed by adenomatosis polyposis coli (APC), a tumor suppressor protein related to APC/beta-catenin signaling pathway. Knockout studies in mice suggested the role of this protein in myelination of the corpus callosum, lipid metabolism, and epidermal cell proliferation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MGC3931; nuclear hormone receptor 1; nuclear receptor subfamily 1 group C member 2; nuclear receptor; PPARdelta; OTTHUMP00000016256; OTTHUMP00000016257; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activator receptor beta; peroxisome proliferator activator receptor, delta; Peroxisome proliferator-activated receptor beta; PPAR-beta; PPAR-delta; PPARd; PPARD/MYO1D fusion; PPARdelta/beta; unnamed protein product

          View more View less

          Gene Aliases: FAAR; NR1C2; NUC-1; NUC1; NUCI; NUCII; PPAR-beta; PPAR-delta; PPAR[b]; PPARB; Pparb/d; PPARdelta

          View more View less

          UniProt ID: (Human) Q03181, (Mouse) P35396

          View more View less

          Entrez Gene ID: (Human) 5467, (Rat) 25682, (Mouse) 19015

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription coactivator binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding steroid hormone receptor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding fatty acid binding protein binding zinc ion binding lipid binding sequence-specific DNA binding NF-kappaB binding linoleic acid binding sequence-specific double-stranded DNA binding signaling receptor activity enzyme activator activity prostacyclin receptor activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter placenta development proteoglycan metabolic process regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter lipid metabolic process fatty acid metabolic process fatty acid beta-oxidation apoptotic process response to stress signal transduction heart development embryo implantation positive regulation of cell proliferation axon ensheathment phospholipid biosynthetic process response to glucose hormone-mediated signaling pathway negative regulation of cardiac muscle hypertrophy positive regulation of gene expression negative regulation of cholesterol storage response to activity regulation of skeletal muscle satellite cell proliferation negative regulation of smooth muscle cell migration fatty acid transport fatty acid oxidation cell differentiation negative regulation of cell growth intracellular receptor signaling pathway cell-substrate adhesion cellular response to nutrient levels negative regulation of collagen biosynthetic process response to vitamin A positive regulation of insulin secretion involved in cellular response to glucose stimulus wound healing regulation of cell proliferation vasodilation negative regulation of apoptotic process positive regulation of skeletal muscle tissue regeneration keratinocyte proliferation regulation of fat cell differentiation negative regulation of myoblast differentiation positive regulation of epidermis development positive regulation of transcription, DNA-templated positive regulation of fatty acid metabolic process positive regulation of transcription from RNA polymerase II promoter positive regulation of fatty acid oxidation decidualization negative regulation of smooth muscle cell proliferation negative regulation of epithelial cell proliferation negative regulation of inflammatory response keratinocyte migration positive regulation of protein kinase B signaling adipose tissue development cellular response to lipopolysaccharide cellular response to hypoxia energy homeostasis apoptotic signaling pathway negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of myoblast proliferation G-protein coupled receptor signaling pathway epidermis development regulation of insulin secretion glucose metabolic process generation of precursor metabolites and energy cholesterol metabolic process cell proliferation fatty acid catabolic process intracellular steroid hormone receptor signaling pathway positive regulation of fat cell differentiation negative regulation of transcription, DNA-templated glucose transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0