Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
PA1-863 detects histone deacetylase 4 (HDAC4) from human cells.
PA1-863 has been successfully used in Western blot and ChIP procedures. By Western blot, this antibody detects an ~140 kDa protein representing HDAC4 from HeLa cell lysate. An ~110 kDa protein is also detected with PA1-863 which could correspond to protein degradation product.
PA1-863 immunogen is a synthetic peptide corresponding to residues M(1)SSQSHPDGLSGRDQPVE(18) of human HDAC4.
PA1-863 can be used with blocking peptide PEP-141 and blocking peptide PEP-095.
Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp779K1053; HD7; HD7b; HD9; HDAC; HDAC4; HDAC7B; HDAC9B; HDAC9FL; HDRP; histone deacetylase A; KIAA0744; MITR; unnamed protein product
Gene Aliases: AHO3; BDMR; HA6116; HD4; HDAC-4; HDAC-A; HDACA; NEDCHF; NEDCHID
UniProt ID: (Human) P56524
Entrez Gene ID: (Human) 9759
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support