Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Calsequestrin Antibodies

          Invitrogen

          Calsequestrin Polyclonal Antibody

          Advanced Verification
          77 Published Figures
          150 References
          View all (21) Calsequestrin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Calsequestrin Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          • Published Figures (77)
          • Advanced Verification (1)
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 84

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calsequestrin Antibody (PA1-913) in ICC/IF

          Immunofluorescent analysis of Calsequestrin (green) showing staining in the cytoplasm and nucleus of C2C12 cells (right) compared to a negative control without primary antibody (left). Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a Calsequestrin polyclonal antibody (Product # PA1-... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Calsequestrin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calsequestrin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calsequestrin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Calsequestrin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Calsequestrin Antibody in Immunocytochemistry (ICC/IF)
          Calsequestrin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calsequestrin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calsequestrin Antibody in Immunohistochemistry (IHC)
          Calsequestrin Antibody in Immunohistochemistry (IHC)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody in Western Blot (WB)
          Calsequestrin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Calsequestrin Polyclonal Antibody

          Product Details

          PA1-913

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:10,000
          View 137 publications 137 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Sheep, Rat

          Published species

          Cat, Dog, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Purified canine cardiac calsequestrin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum, PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2071461

          Product Specific Information

          PA1-913 detects calsequestrin from canine, human, mouse, rat and sheep tissues. This antibody recognizes both cardiac and skeletal muscle calsequestrin.

          PA1-913 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, immunohistochemistry (paraffin) and immunoprecipitation procedures. By Western blot, this antibody detects an ~55 kDa protein representing calsequestrin from canine cardiac extract. Additional bands at 97 kDa may be observed and have been reported to be calsequestrin-like proteins.

          PA1-913 antigen is purified canine cardiac calsequestrin.

          Target Information

          The sarcoplasmic reticulum (SR) is, in part, responsible for maintaining the level of intracellular calcium in cardiac and skeletal muscle by storing and releasing calcium. Several intralumenal SR calcium binding proteins have been identified, the most prominent of these is calsequestrin. Calsequstrin is a calcium binding protein known to sequester calcium accumulated in the sarcoplasmic reticulum of muscle cells during relaxation and is found discretely localized to the junctional and corbular (terminal cisternae) SR. Calsequestrin functions to localize calcium near the junctional face of the terminal cisternae from which calcium can be released into the cytosol via the ryanodine receptor. This protein is highly acidic and has a large capacity and moderate to low affinity for calcium.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Aspartactin; calmitin; calmitine; calsequestrin 1 (fast-twitch, skeletal muscle); Calsequestrin-1; Laminin-binding protein; mitochondrial calcium-binding protein; unnamed protein product

          View more View less

          Gene Aliases: CASQ; CSQ; CSQ-1; CSQ1; PDIB1; sCSQ; VMCQA

          View more View less

          UniProt ID: (Human) P31415, (Rat) P19633, (Mouse) O09165

          View more View less

          Entrez Gene ID: (Sheep) 101118652, (Dog) 608401, (Human) 844, (Rat) 686019, (Mouse) 12372

          View more View less

          Function(s)
          calcium ion binding identical protein binding protein binding calcium-binding protein
          Process(es)
          positive regulation of release of sequestered calcium ion into cytosol positive regulation of store-operated calcium channel activity regulation of store-operated calcium entry skeletal muscle tissue development regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion response to muscle inactivity response to denervation involved in regulation of muscle adaptation sarcomere organization protein polymerization regulation of release of sequestered calcium ion into cytosol endoplasmic reticulum organization response to heat regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of muscle system process regulation of muscle contraction regulation of striated muscle contraction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0