Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • eNOS Antibodies

          Invitrogen

          eNOS Polyclonal Antibody

          9 Published Figures
          29 References
          View all (44) eNOS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite eNOS Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (9)
          eNOS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          eNOS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          eNOS Antibody (PA3-031A) in ICC/IF

          Immunofluorescent analysis of eNOS using eNOS Polyclonal Antibody (Product # PA3-031A) shows staining in A2058 Cells. eNOS (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with an antibody recognizing eNOS (Product # PA3-031A) at a dilution of 1:200 over night at 4 °C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35552 for GAR, Product # 35... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          eNOS Antibody in Immunocytochemistry (ICC/IF)
          eNOS Antibody in Immunocytochemistry (ICC/IF)
          eNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          eNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          eNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          eNOS Antibody in Immunohistochemistry (IHC)
          eNOS Antibody in Immunohistochemistry (IHC)
          eNOS Antibody in Western Blot (WB)
          eNOS Antibody in Immunocytochemistry (ICC/IF)
          eNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          eNOS Antibody in Immunohistochemistry (IHC)
          eNOS Antibody in Immunohistochemistry (IHC)
          eNOS Antibody in Immunohistochemistry (IHC)
          eNOS Antibody in Immunohistochemistry (IHC)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          eNOS Polyclonal Antibody

          Product Details

          PA3-031A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 16 publications 16 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:250
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          1:250
          View 5 publications 5 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Dog, Human, Mouse, Rat

          Published species

          Bovine, Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues P(599) Y N S S P R P E Q H K S Y K(613) C of bovine eNOS.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum, PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2251447

          Product Specific Information

          PA3-031A detects endothelial Nitric Oxide Synthase (eNOS) from canine, bovine, human, mouse and rat tissues. This antibody does not detect brain NOS (bNOS) or inducible NOS (iNOS).

          PA3-031A has been successfully used in Western blot, immunohistochemistry, immunofluorescence, and immunocytochemistry procedures. By Western blot, this antibody detects an approximately 140 kDa band representing eNOS from canine cardiac extracts. Immunohistochemical staining of eNOS in mouse cardiac tissue with PA3-031 results in the staining of the endothelium.

          The PA3-031A immunogen is a synthetic peptide corresponding to residues P(599) Y N S S P R P E Q H K S Y K(613) C of bovine eNOS.

          Target Information

          Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) and endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cNOS; constitutive endothelial nitric oxide synthase; constitutive NOS; e-NOS; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial NOS; eNOS; nitric oxide synthase; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS type III; Nos3; NOSIII; OTTHUMP00000213183; unnamed protein product

          View more View less

          Gene Aliases: 2310065A03Rik; cNOS; EC-NOS; ECNOS; eNOS; MYMY8; NOS; Nos-3; NOSIII

          View more View less

          UniProt ID: (Human) P29474, (Mouse) P70313, (Rat) Q62600

          View more View less

          Entrez Gene ID: (Bovine) 287024, (Dog) 403784, (Human) 4846, (Mouse) 18127, (Rat) 24600

          View more View less

          Function(s)
          nitric-oxide synthase activity calmodulin binding FMN binding heme binding metal ion binding flavin adenine dinucleotide binding NADP binding actin monomer binding protein binding superoxide-generating NADPH oxidase activity oxidoreductase activity tetrahydrobiopterin binding arginine binding cadmium ion binding scaffold protein binding actin binding beta-catenin binding cadherin binding nitric-oxide synthase binding Hsp90 protein binding oxidoreductase protein modifying enzyme
          Process(es)
          arginine catabolic process nitric oxide biosynthetic process mitochondrion organization blood coagulation positive regulation of guanylate cyclase activity oxidation-reduction process cellular response to laminar fluid shear stress negative regulation of extrinsic apoptotic signaling pathway via death domain receptors negative regulation of leukocyte cell-cell adhesion blood vessel remodeling regulation of systemic arterial blood pressure by endothelin aortic valve morphogenesis pulmonary valve morphogenesis endocardial cushion morphogenesis blood circulation regulation of blood pressure negative regulation of cell proliferation response to heat response to hormone negative regulation of platelet activation positive regulation of gene expression negative regulation of muscle hyperplasia smooth muscle hyperplasia removal of superoxide radicals regulation of neurological system process response to lipopolysaccharide response to fluid shear stress nitric oxide-cGMP-mediated signaling pathway vasodilation positive regulation of blood vessel endothelial cell migration endothelial cell migration cell redox homeostasis positive regulation of Notch signaling pathway positive regulation of angiogenesis negative regulation of blood pressure tetrahydrobiopterin metabolic process nitric oxide metabolic process homeostasis of number of cells within a tissue ventricular septum morphogenesis negative regulation of biomineral tissue development regulation of blood vessel diameter angiogenesis ovulation from ovarian follicle in utero embryonic development regulation of the force of heart contraction regulation of sodium ion transport regulation of the force of heart contraction by chemical signal protein import into nucleus gene expression lung development lipopolysaccharide-mediated signaling pathway response to estradiol positive regulation of apoptotic process negative regulation of vascular permeability negative regulation of potassium ion transport negative regulation of smooth muscle cell proliferation negative regulation of hydrolase activity negative regulation of calcium ion transport cellular response to mechanical stimulus cellular response to transforming growth factor beta stimulus endothelial tube formation response to hypoxia female pregnancy response to xenobiotic stimulus response to mechanical stimulus response to carbohydrate response to fructose response to metal ion response to lead ion response to activity response to nutrient levels cellular response to insulin stimulus response to cytokine cellular response to heat response to nicotine response to erythropoietin response to decreased oxygen levels regulation of apoptotic process response to amino acid response to peptide hormone response to ethanol response to mercury ion response to axon injury response to corticosterone response to hyperoxia retina development in camera-type eye bone development response to magnetism cellular response to magnesium ion cellular response to cAMP cellular response to tumor necrosis factor cellular response to growth factor stimulus cellular response to high density lipoprotein particle stimulus cellular response to low-density lipoprotein particle stimulus response to peptide response to ketone response to salt response to L-arginine cellular response to rotenone response to ultrasound
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch