Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KLLEAISETS SQLEHAKVTQ TELMRESFRQ KQEATESLKC QEELRERLHE ESRAREQLAV ELSKAEGVID GYADEKTLFE RQIQEKTDII DRLEQELLCA SNRLQELEAE QQQIQEEREL LSRQKEAMKA EAGPVEQQLL QETE
Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 86%.
The A-kinase anchor proteins are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A kinase (PRKA) anchor protein (yotiao) 9; A kinase (PRKA) anchor protein 9; A-kinase anchor protein 350 kDa; A-kinase anchor protein 450 kDa; A-kinase anchoring protein; putative centrosomally assiciated AKAP; AKAP 120-like protein; AKAP9-BRAF fusion protein; centrosomally-associated AKAP; centrosome- and Golgi-localized PKN-associated protein; centrosome- and golgi-localized protein kinase N-associated protein; hypothetical; AKAP450; kinase N-associated protein; protein hyperion; protein kinase A anchoring protein 9; protein phosphatase 1, regulatory subunit 45; protein yotiao; unnamed protein product
Gene Aliases: AKAP-9; AKAP350; AKAP450; CG-NAP; HYPERION; LQT11; MU-RMS-40.16A; PPP1R45; PRKA9; YOTIAO
UniProt ID: (Human) Q99996
Entrez Gene ID: (Human) 10142
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support