Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: EAVCQDVLSL LDNYLIKNCS ETQYESKVFY LKMKGDYYRY LAEVATGEKR ATVVESSEKA YSEAHEIS
Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
14-3-3 gamma encodes a product that belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the rat ortholog. It is induced by growth factors in human vascular smooth muscle cells, and is also highly expressed in skeletal and heart muscles, suggesting an important role for this protein in muscle tissue. It has been shown to interact with RAF1 and protein kinase C, proteins involved in various signal transduction pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KCIP-1; protein kinase C inhibitor protein 1; protein phosphatase 1, regulatory subunit 170; signal transduction phosphoprotein; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; unnamed protein product; YWHAG
Gene Aliases: 14-3-3GAMMA; DEE56; EIEE56; PPP1R170
UniProt ID: (Human) P61981
Entrez Gene ID: (Human) 7532
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support