Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LHHALHEDGH EAQLEPSPPP HLGAHGHAHG HAHAG
Highest antigen sequence indentity to the following orthologs: Rat - 40%, Mouse - 100%.
POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-Oct; oct-6; octamer-binding protein 6; octamer-binding transcription factor 6; OTF-6; POU domain transcription factor SCIP
Gene Aliases: OCT6; OTF6; SCIP
UniProt ID: (Human) Q03052
Entrez Gene ID: (Human) 5453
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support