Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.
Immunogen sequence: METKKLIGKP LQPARPVRHL TSPPGAVFPF NFQNEYPCNT QCIQSGVSRC K
Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 29%.
Predicted to enable eukaryotic initiation factor 4E binding activity. Predicted to be involved in negative regulation of translational initiation. Predicted to be active in cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chromosome 8 open reading frame 88
UniProt ID: (Human) P0DMB2
Entrez Gene ID: (Human) 100127983
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support