Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LPYAMKPIDY YTETKILQER AVLGANDPEK NFLTTAIRPH GIFGPRDPQL VPILIEAARN GKMKFVIGNG KNLVDFTFVE NVVHGHILAA EQLSRDSTLG GKAFHITNDE PIPFWTFLSR I
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-beta-hydroxysteroid dehydrogenase; 3beta-hydroxysteroid-4alpha-carboxylate 3-dehydrogenase (decarboxylating); bare patches; epididymis secretory sperm binding protein; H105e3; NAD(P) dependent steroid dehydrogenase-like; NAD(P) dependent steroid dehydrogenase-like protein transcript; NAD(P)-dependent steroid dehydrogenase-like protein; protein H105e3; short chain dehydrogenase/reductase family 31E, member 1; striated; unnamed protein product
Gene Aliases: Bpa; H105E3; SDR31E1; Str; XAP104
UniProt ID: (Human) Q15738, (Rat) Q5PPL3, (Mouse) Q9R1J0
Entrez Gene ID: (Human) 50814, (Rat) 309262, (Mouse) 18194
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support