Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAASVKEQST KPIPLPQSTP GEGDNDEEDP SKLKEEQHGI SVTGLQSPDR DLGLESTLIS SKPQSHSLST SGKSEVRDLF VAERQFAKEQ HTDGTLKEVG EDYQ
Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli working through G protein-coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein has been observed to form a myeloid/lymphoid fusion partner in acute myeloid leukemia. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KMT2A/ARHGEF12 fusion protein; leukemia-associated rho guanine nucleotide exchange factor; leukemia-associated RhoGEF; Rho guanine nucleotide exchange factor (GEF) 12; unnamed protein product
Gene Aliases: LARG; PRO2792
UniProt ID: (Human) Q9NZN5
Entrez Gene ID: (Human) 23365
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support