Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: HSLAKSLCIL NISNNNIDDI TDLELLENLN QLIAVDNQLL HVKDLEFLLN KLMKLWKIDL NGNPVCLKPK YRDRLILVSK SLGT
Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 67%.
Regulates phosphatase activity of protein phosphatase 1 (PP1) complexes in the testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine rich repeat containing 67; leucine-rich repeat-containing protein 67; testis leucine-rich repeat
Gene Aliases: dtr; LRRC67; TLLR; TLRR
UniProt ID: (Human) Q7Z4L9
Entrez Gene ID: (Human) 286187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support