Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ILVDSGMKEN GKVISFFKLK ESQLPALAIY QTLDDEWDTL PTAEVSVEHV QNFCDGFLSG KLLKENRESE GKTPKV
Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 57%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum protein 27 kDa; ER protein 27; inactive protein disulfide-isomerase 27; protein disulfide isomerase family A, member 8; unnamed protein product
Gene Aliases: C12orf46; PDIA8
UniProt ID: (Human) Q96DN0
Entrez Gene ID: (Human) 121506
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support