Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SLSEDPCPSQ KALDPAPLAR PSPAGSAQTS PELEHRVSLF NQKNQEGFTV FQIRPVIHFQ PTVPMLEDKF RSLESKEQKL HRV
Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
C15orf27 is a voltage-sensor protein present on the post-synaptic side of glutamatergic mossy fibers and granule cells in the cerebellum. Despite the presence of a voltage-sensor segment, it does not form a functional ion channel and its precise role remains unclear. The protein undergoes both rapid and slow structural rearrangements in response to changes in voltage, and contains a zinc-binding site that can regulate the slow conformational transition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hv1 related protein 1; transmembrane protein C15orf27; unnamed protein product
Gene Aliases: C15orf27; HsHVRP1; hTMEM266; HVRP1
UniProt ID: (Human) Q2M3C6
Entrez Gene ID: (Human) 123591
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support