Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SSSGSSSAHS NQQQPPGSVS QEPRPPLQQS QVPPPEVRMT VPPLATSSAP VAVPSTAPVT YPMPQTPMGC PQPTPKMETP AIRPPPHGTT APHKN
Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
This gene encodes a protein with ankyrin repeats, which are associated with protein-protein interactions. Studies in mice suggest that this protein is involved in liver development. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene trap ankyrin repeat protein; serologically defined breast cancer antigen NY-BR-16; unnamed protein product
Gene Aliases: CAGS; GTAR; MASK2; NY-BR-16
UniProt ID: (Human) O75179
Entrez Gene ID: (Human) 26057
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support