Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GTGTLLITLE DVNDNAPFIY PTVAEVCDDA KNLSVVILGA SDKDLHPNTD PFKFEIHKQA VPDKVWKISK INNTHALVSL LQNLNKANYN LPIMVTDSGK PPMTNITDLR VQVCSCRNSK VDCNAAGALR FSLPSVLLLS LFSLA
Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.
The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAD13; cadherin 13, H-cadherin (heart); CD39 antigen-like 3; CD39-like 3; CD39L3; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; H-cadherin (heart); HB6; heart cadherin; heart-cadherin; NTPDase 3; NTPDase-3; T cad; T Cadherin; T-cad; T-cadherin; truncated-cadherin; unnamed protein product
Gene Aliases: CDHH; P105
UniProt ID: (Human) P55290
Entrez Gene ID: (Human) 1012
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support