Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GVGGDPAQLA QRISSNIQKI TQCSVEIQRT LNQLGTPQDS PELRQQLQQK QQYTNQLAKE TDKYIKEFGS LPTTPSEQRQ RKIQKDRLVA EFTTSLTNFQ KVQRQAAERE KEFVARVRAS SRVSGSFPED SSKERNLVSW ESQTQPQV
Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 94%.
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: member of the syntaxin family; syntaxin-7-like; Syntaxin7; t-SNARE; unnamed protein product
Gene Aliases: Syn7
UniProt ID: (Human) O15400, (Rat) O70257
Entrez Gene ID: (Human) 8417, (Mouse) 53331, (Rat) 60466
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support