Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TEELHSAQQW NGQGILELLR TVPIAGGPWL ITDMRRGETI FEIDPHLQER VDKGIETDGS NLSGVSAKCA WDDLSRPPED DEDSRSICIG TQPRRLSGKD TEQIRETLRR GLEINSKPVL PPINPQRQNG LAHDRAPSRK DSLESDSSTA
Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 97%.
The Hedgehog signaling pathway plays an important role in early human development. The pathway is a signaling cascade that plays a role in pattern formation and cellular proliferation during development. This gene encodes a negative regulator of the hedgehog signaling pathway. Defects in this gene are a cause of medulloblastoma. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: negative regulator of hedgehog signaling; novel; novel PEST-containing protein; novel; su(fu); OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; RP11-47A8.1; unnamed protein product
Gene Aliases: BCNS2; JBTS32; PRO1280; SUFUH; SUFUXL
UniProt ID: (Human) Q9UMX1
Entrez Gene ID: (Human) 51684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support