Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SLHSIAVSSI QKAKGYQLLE EEKIVSHYFP LTEVGMVETS PLPAKPFNQF EEKISTGPLG DLSRGYQETL PSYAQVGAQE VEGEGPPAEE GAEPEVGEKK EEAERLTTEE QEKVAVPEGE KVETPGVDKE GEKEEPQSEK VSKQGLP
Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 81%.
This gene encodes a transmembrane connexin protein that is necessary for lens growth and maturation of lens fiber cells. The encoded protein is a component of gap junction channels and functions in a calcium and pH-dependent manner. Mutations in this gene have been associated with zonular pulverulent cataracts, nuclear progressive cataracts, and cataract-microcornea syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell surface glycoprotein; connexin 50; gap junction alpha 8; gap junction membrane channel protein alpha-8; gap junction protein alpha 8 50kDa; lens fiber protein MP70; lens intrinsic membrane protein MP70
Gene Aliases: CAE; CAE1; CTRCT1; CX50; CZP1; MP70
UniProt ID: (Human) P48165
Entrez Gene ID: (Human) 2703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support