Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ENWRVDSAAT SGYEIGNPPD YRGQSCMKRH GIPMSHVARQ ITKEDFATFD YILCMDESNL RDLNRKSNQV KTCKAKIELL GSYDPQKQLI IEDPYYGNDS DFETVYQ
Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.
Low molecular weight protein tyrosine phosphtases, including the human red cell phosphatase, HCPTP, are widely expressed cytosolic proteins of approximately 18 kDa that exist in distinct isoforms and are highly selective for phosphotyrosine over phosphoserine or phosphotyrosine. The HCPTP-A and HCPTP-B proteins expressed in human red cells and placenta are fast and slow forms of red cell acid phosphatase5. Like other tyrosine phosphatases, HCPTP is sensitive to inhibition by vanadate, and has a catalytic mechanism that involves formation of a cystinylphosphate intermediate6. The crystal structure of the bovine heart derived enzyme BHPTP shows a four-stranded central parallel b sheet with flanking a helices and an active site cysteine residue in the typical tyrosine phosphatase sequence context, CXXXXXR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid phosphatase 1, soluble; acid phosphatase of erythrocyte; adipocyte acid phosphatase; cytoplasmic phosphotyrosyl protein phosphatase; HAAP betaÊ; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; LMW-PTPase; low molecular weight cytosolic acid phosphatase; MGC111030; MGC3499; protein tyrosine phosphatase; red cell acid phosphatase 1; testicular secretory protein Li 37; unnamed protein product
Gene Aliases: 4632432E04Rik; Acp-1; HAAP; Lmptp; LMW-PTP; LMWPTP
UniProt ID: (Human) P24666, (Rat) P41498, (Mouse) Q9D358
Entrez Gene ID: (Human) 52, (Rat) 24161, (Mouse) 11431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support