Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: IHLLDLKDRS SIENLWGLQP RPPASLLQPT ASYSRKDKDQ RKQQAMWRVP SDLKMLKRLK TQMAEVRCMK TDVKNTLSEI KSSSAASGDM QTSLFSADQA ALAACGTENS GRLQDLGMEL LAKSS
Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.
Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mulibrey nanism protein; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; unnamed protein product
Gene Aliases: MUL; POB1; TEF3
UniProt ID: (Human) O94972
Entrez Gene ID: (Human) 4591
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support