Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SLLHCFQCCG AKVLLVSPEL QAAVEEILPS LKKDDVSIYY VSRTSNTDGI DSFLDKVDEV STEPIPESWR SEVTFSTPAL YIYTSGTTGL PKAAMITHQR IWYGTGLTFV SGLKADDVIY ITLPFYHSAA LLIGIHGCIV AGAT
Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 73%.
The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arachidonate--CoA ligase; FATP-2; fatty acid transport protein 2; fatty-acid-coenzyme A ligase, very long-chain 1; long-chain-fatty-acid--CoA ligase; phytanate--CoA ligase; solute carrier family 27 (fatty acid transporter), member 2; THCA-CoA ligase; unnamed protein product; ve; very long-chain acyl-CoA synthetase; very long-chain fatty-acid-coenzyme A ligase 1; very long-chain-fatty-acid-CoA ligase
Gene Aliases: ACSVL1; FACVL1; FATP2; hFACVL1; HsT17226; VLACS; VLCS
UniProt ID: (Human) O14975
Entrez Gene ID: (Human) 11001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support