Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LVELQEQLQQ LPALIADLES MTANLTHLEA SFEEVENNLL HLEDLCGQCE LERCKHMQSQ QLENYKK
Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 73%.
Dtnbp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 8; biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; DKFZp564K192; DTBP1; dysbindin-1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; Hps7-like protein; MGC20210; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1; subunit of BLOC-1; unnamed protein product
Gene Aliases: BLOC1S8; DBND; HPS7; My031; SDY
UniProt ID: (Human) Q96EV8
Entrez Gene ID: (Human) 84062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support