Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TEADVNPKAY PLADAHLTKK LLDLVQQSCN YKQLRKGANE ATKTLNRGIS EFIVMAADAE PLEIILHLPL LCEDKNVPYV FVRSKQALGR ACGVSRPVIA CSVTIKEGSQ LKQQIQSIQQ SIERL
Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; CTA-216E10.8; high mobility group-like nuclear protein 2 homolog 1; NHP2 non-histone chromosome protein 2-like 1; NHP2L1; non-histone chromosome protein 2-like 1; SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5); sperm specific antigen 1; U4/U6.U5 tri-snRNP 15.5 kDa protein; unnamed protein product
Gene Aliases: 15.5K; FA-1; FA1; NHP2L1; NHPX; OTK27; SNRNP15-5; SPAG12; SSFA1
UniProt ID: (Human) P55769
Entrez Gene ID: (Human) 4809
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support