Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KDTDKFKGFC YVEFDEVDSL KEALTYDGAL LGDRSLRVDI AEGRKQDKGG FGFRKGGPDD RGMGSSRESR GGWDSRDDFN SG
Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 99%.
This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4H; KIAA0038; unnamed protein product; williams-Beuren syndrome chromosomal region 1 protein homolog; Williams-Beuren syndrome chromosome region 1; Williams-Beuren syndrome chromosome region 1 homolog
Gene Aliases: D5Ertd355e; E430026L18Rik; Ef4h; eIF-4H; WBSCR1; WSCR1
UniProt ID: (Human) Q15056, (Mouse) Q9WUK2, (Rat) Q5XI72
Entrez Gene ID: (Human) 7458, (Mouse) 22384, (Rat) 288599
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support