Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: AFPPREYWSE PYPLPPPTSV LQEPPVQSPG AGRSPWGRAG SLAKEQASVY TKLCGVFPPH LVEAVMGRFP QLLDPQQLAA EILSYKSQHP S
Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 84%.
ZC3H12A is an MCP1 -induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bifunctional endoribonuclease and deubiquitinase ZC3H12A; FLJ23231; MCP-1 induced protein 1; MCPIP1; MCP-1 treatment-induced protein; MCP-induced protein 1; MCPIP; transcription factor; monocyte chemotactic protein-induced protein 1; regnase-1; ribonuclease ZC3H12A; RP3-423B22.1; unnamed protein product; zinc finger CCCH domain-containing protein 12A
Gene Aliases: dJ423B22.1; MCPIP; MCPIP-1; MCPIP1; Reg1
UniProt ID: (Human) Q5D1E8
Entrez Gene ID: (Human) 80149
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support