Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: FLDFSDKEAN TAMGLFTDLA LEERFQVWFQ ANLPGVLPAL TGVLKTDPKV SSSSALCQCI AIMGNLSAEP TTRRHMAACE EFGDGC
Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 81%.
TTC12 contains 3 TPR repeats. Hypermethylation of TTC12 gene may play a role in acute lymphoblastic leukemia. Haplotypic variants in DRD2, ANKK1, TTC12, and NCAM1 are associated with comorbid alcohol and drug dependence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tetratricopeptide repeat and armadillo repeat protein; TPR repeat protein 12; unnamed protein product
Gene Aliases: CILD45; TPARM
UniProt ID: (Human) Q9H892
Entrez Gene ID: (Human) 54970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support