Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ASTMKEVVYW SPKKVADWLL ENAMPEYCEP LEHFTGQDLI NLTQEDFKKP PLCRVSSDNG QRLLDMIETL KMEHHLEAHK NGHANGHLNI GVDIPTPDGS FSIKIKPNGM PNGYRKEMIK IPM
Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: medulla oblongata-derived protein; MGC17342; protein Mob; putative transmembrane protein; transmembrane protein; transmembrane protein 23; unnamed protein product
Gene Aliases: hmob33; MOB; MOB1; SMS1; TMEM23
UniProt ID: (Human) Q86VZ5
Entrez Gene ID: (Human) 259230
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support