Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: WGCLRNPHSE GNGASGLPHY PPTPFSFHQK PDFLATATAA YPDFSASCLA ATPHSLPQEE HIFTEQHPAF PQSPNWHFPV SDARRRPNSG PAGGSKEMGT SSLGLVDTTG GPGDDYGVLG STANETEKKS SR
Highest antigen sequence indentity to the following orthologs: Mouse - 74%, Rat - 74%.
MEOX1 belongs to a family of nonclustered, diverged homeobox genes. It may play a role in regulating growth and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GP91-2; mitogenic oxidase 1; NOH-1; NOH1; NOX-1; unnamed protein product
Gene Aliases: KFS2; MOX1
UniProt ID: (Human) P50221
Entrez Gene ID: (Human) 4222
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support