Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: QFATNGEKSL LFDAMNMTWT VINHEASKIK ETWKKDRGLE KYFRKLSKGD CDHWLREFLG HWEAMPEPTV SPVNASDIHW SSSSLP
Highest antigen sequence indentity to the following orthologs: Rat - 30%, Mouse - 28%.
This gene belong to the RAET1 family, which consists of major histocompatibility complex (MHC) class I-related genes located in a cluster on chromosome 6q24.2-q25.3. This and RAET1G protein differ from other RAET1 proteins in that they have type I membrane-spanning sequences at their C termini rather than glycosylphosphatidylinositol anchor sequences. This protein functions as a ligand for NKG2D receptor, which is expressed on the surface of several types of immune cells, and is involved in innate and adaptive immune responses. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Aug 2011].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ligand for NKG2D receptor; lymphocyte effector toxicity activation ligand; MGC125309; NKG2D ligand 4; NKG2D-ligand; RAE-1-like transcript 4; RAE1 like transcript 4; RL-4; membrane glycoprotein; stress-inducible antigen; UL16 binding protein 4; UL16-binding protein 4; ULBP 4; ULBP4MGC125308
Gene Aliases: bA350J20.7; LETAL; N2DL-4; NKG2DL4; RAET1E2; RL-4; ULBP4
UniProt ID: (Human) Q8TD07
Entrez Gene ID: (Human) 135250
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support