Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: AKSQEDTEAV FKKALLAVIG ADSVTLKNKY LDWAYRSGGY KKARAVFKSL QESRPFSVDF FRKMIQFEKE QESCNMANIR EYYERALREF GSADSDLWMD YMKEELNHPL GRPENCGQIY WRAMKMLQG
Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 87%.
Involved in nucleolar processing of pre-18S ribosomal RNA. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hepatocellular carcinoma-associated antigen 66; multiple hat domains protein; predicted protein of HQ1289; unnamed protein product; UTP6, small subunit (SSU) processome component, homolog
Gene Aliases: C17orf40; HCA66
UniProt ID: (Human) Q9NYH9
Entrez Gene ID: (Human) 55813
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support