Thermo Fisher Scientific Logo
๋กœ๊ทธ์ธ
ํšŒ์›์ด ์•„๋‹ˆ์‹ ๊ฐ€์š”? ๊ณ„์ • ์ƒ์„ฑํ•˜๊ธฐ
  • ๋ชจ๋“  ์ œํ’ˆ ์ฃผ๋ฌธ
    • ํ•ญ์ฒด
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR ๋ถ„์„
    • ํ”ผํŽซ ๋ฐ ํ”ผํŽซ ํŒ
    • ์‹คํ—˜์‹ค์šฉ ์›์‹ฌ๋ถ„๋ฆฌ๊ธฐ
    • ์ดˆ์ €์˜จ ๋ƒ‰๋™๊ณ 
    • ๋ถ„๊ด‘ํ•™
    • ๋น„์ด์ปค
    • PCR ์žฅ๋น„ ๋ฐ ์šฉํ’ˆ
    • ์ œํ’ˆ ์นดํ…Œ๊ณ ๋ฆฌ ์ „์ฒด๋ณด๊ธฐ
  • ์‘์šฉ ๋ถ„์•ผ ๋ฐ ๊ธฐ๋ฒ•
    • ์„ธํฌ ๋ถ„์„
    • ์‹คํ—˜์‹ค ์žฅ๋น„
    • Real-Time PCR
    • PCR
    • ํฌ๋กœ๋งˆํ† ๊ทธ๋ž˜ํ”ผ
    • ์„ธํฌ ๋ฐฐ์–‘ ๋ฐ ํŠธ๋žœ์ŠคํŽ™์…˜
    • DNA/RNA ์ถ”์ถœ๊ณผ ๋ถ„์„
    • ๋‹จ๋ฐฑ์งˆ ์ƒ๋ฌผํ•™
    • ์œ ์„ธํฌ๋ถ„์„
    • ํ™”ํ•™ ์ œํ’ˆ
    • ์‘์šฉ ๋ถ„์•ผ ๋ฐ ๊ธฐ๋ฒ• ์ „์ฒด๋ณด๊ธฐ
  • ์„œ๋น„์Šค
    • ๋งž์ถคํ˜• ์„œ๋น„์Šค
    • ์—”ํ„ฐํ”„๋ผ์ด์ฆˆ๊ธ‰ ์‹คํ—˜์‹ค ์ •๋ณด ์ฒ˜๋ฆฌ
    • 360ยฐ CDMO ๋ฐ CRO ์„œ๋น„์Šค
    • CDMO ์„œ๋น„์Šค
    • ์ž„์ƒ์‹œํ—˜ CRO ์„œ๋น„์Šค
    • ์žฅ๋น„ ์„œ๋น„์Šค
    • ๊ต์œก ์„œ๋น„์Šค
    • Unity Lab Services
    • ์„œ๋น„์Šค ์ „์ฒด ๋ณด๊ธฐ
  • ์ง€์›
    • ๊ณ ๊ฐ์„ผํ„ฐ
    • ๋ฌธ์˜ํ•˜๊ธฐ
    • ์‹œํ—˜์„ฑ์ ์„œ(COA) ๋ฐ ์ ํ•ฉ์„ฑ ์ธ์ฆ์„œ(COC)
    • Safety Data Sheets (SDS)
    • ๋งค๋‰ด์–ผ
    • ์ธ์šฉ ๋ฐ ์ฐธ๊ณ  ๋ฌธํ—Œ
    • ๊ธฐ๊ธฐ ์ง€์›
    • ๊ธฐ์ˆ  ์ž๋ฃŒ/์ œํ’ˆ FAQ
    • ํ•™์Šต ์„ผํ„ฐ
    • ์ง€์› ๋ฉ”๋‰ด ์ „์ฒด๋ณด๊ธฐ
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • ์‹ ์ œํ’ˆ
  • ๋ฌธ์˜ํ•˜๊ธฐ
  • ๋น ๋ฅธ ์ฃผ๋ฌธ
  • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
  • ์ œํ’ˆ ๋ฌธ์„œ ๊ฒ€์ƒ‰
Thermo Fisher Scientific Logo

Search

์ „์ฒด๊ฒ€์ƒ‰
          • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
          • ๋น ๋ฅธ์ฃผ๋ฌธ
          • ๋กœ๊ทธ์ธ
            ๋กœ๊ทธ์ธ
            ํšŒ์›์ด ์•„๋‹ˆ์‹ ๊ฐ€์š”? ๊ณ„์ • ์ƒ์„ฑํ•˜๊ธฐ
            • ๊ณ„์ •์ •๋ณด
            • ์ฃผ๋ฌธ๋‚ด์—ญ์กฐํšŒ
            • ์ปค์Šคํ…€์ œํ’ˆ ๋ฐ ํ”„๋กœ์ ํŠธ
            • Services Central
          • Primary Antibodies ›
          • BACH2 Antibodies

          Invitrogen

          BACH2 Polyclonal Antibody

          1 Reference
          View all (8) BACH2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite BACH2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          BACH2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BACH2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BACH2 Antibody (PA5-84376) in ICC/IF

          Immunofluorescent analysis of BACH2 in human cell line SH-SY5Y using BACH2 Polyclonal Antibody (Product # PA5-84376). Staining shows localization to nucleus and cytosol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BACH2 Antibody in Immunocytochemistry (ICC/IF)
          BACH2 Antibody in ChIP Assay (ChIP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as โ€œraw-uneditedโ€, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          BACH2 Polyclonal Antibody

          Product Details

          PA5-84376

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          0.25-2 ยตg/mL
          -

          ChIP assay (ChIP)

          1-10 ยตg
          -
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to Human BACH2. Recombinant protein control fragment (Product #RP-106850).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.05 mg/mL

          Amount

          5 ยตg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 40% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4ยฐC short term. For long term storage, store at -20ยฐC, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2791528

          Product Specific Information

          Immunogen sequence: SNSLKPGLAR GQIKSEPPSE ENEEESITLC LSGDEPDAKD RAGDVEMDRK QPSPAPTPTA PAGAACLERS RSVASPSCLR SLFSITK

          Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 85%.

          Target Information

          BTB and CNC homolog 2 (BACH2) is a transcriptional regulator that acts as repressor or activator. BACH2 binds to Maf recognition elements (MARE). It induces apoptosis in response to oxidative stress through repression of the antiapoptotic factor HMOX1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BTB and CNC homolog 2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; BTB domain and CNC homolog 2

          View more View less

          Gene Aliases: BTBD25; IMD60

          View more View less

          UniProt ID: (Human) Q9BYV9

          View more View less

          Entrez Gene ID: (Human) 60468

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter primary adaptive immune response involving T cells and B cells
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          ์˜จ๋ผ์ธ ์ฃผ๋ฌธ Plus Icon Minus Icon
          • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
          • ์ฃผ๋ฌธ ์ง€์›
          • ๋น ๋ฅธ ์ฃผ๋ฌธ
          • Supply Center
          • eProcurement
          ์ง€์› Plus Icon Minus Icon
          • Help and Support
          • ๊ณ ๊ฐ ์„ผํ„ฐ
          • ๊ธฐ์ˆ  ์ง€์› ์„ผํ„ฐ
          • ์ œํ’ˆ ๋ฌธ์„œ ๊ฒ€์ƒ‰
          • ์‚ฌ์ดํŠธ ๋ฌธ์ œ ๋ณด๊ณ 
          ๊ต์œก ๋ฐ ์ด๋ฒคํŠธ Plus Icon Minus Icon
          • ๊ต์œก ์„ผํ„ฐ
          • ํ”„๋กœ๋ชจ์…˜
          • ์ด๋ฒคํŠธ ๋ฐ ์›จ๋น„๋‚˜
          • ์†Œ์…œ ๋ฏธ๋””์–ด
          About Thermo Fisher Plus Icon Minus Icon
          • ์†Œ๊ฐœ ์†Œ๊ฐœ
          • ์ฑ„์šฉ ์ฑ„์šฉ
          • ํˆฌ์ž์ž ํˆฌ์ž์ž
          • ๋‰ด์Šค ๋‰ด์Šค
          • ์‚ฌํšŒ์  ์ฑ…์ž„ ์‚ฌํšŒ์  ์ฑ…์ž„
          • Trademarks
          • ๊ณต์ •๊ฑฐ๋ž˜ ๊ณต์ •๊ฑฐ๋ž˜
          • ์ •์ฑ… ๋ฐ ๊ณ ์ง€
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • ์ด์šฉ ์•ฝ๊ด€
          • ๊ฐœ์ธ์ •๋ณด์ฒ˜๋ฆฌ๋ฐฉ์นจ
          • ๊ฐ€๊ฒฉ ๋ฐ ์šด์ž„ ์ •์ฑ…
          ยฉ 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด

           

          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat

          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด

           

          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat

          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด

           

          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat

          ์จ๋ชจ ํ”ผ์…” ์‚ฌ์ด์–ธํ‹ฐํ”ฝ ์ฝ”๋ฆฌ์•„ ์ฃผ์‹ํšŒ์‚ฌ
          ๋Œ€ํ‘œ์ž : ์„์ˆ˜์ง„
          ์‚ฌ์—…์ž ๋“ฑ๋ก๋ฒˆํ˜ธ : 117-81-46910
          ์ž…๊ธˆ๊ณ„์ขŒ : ํ•˜๋‚˜์€ํ–‰ 336-890014-06204
          (์˜ˆ๊ธˆ์ฃผ : ์จ๋ชจํ”ผ์…”์‚ฌ์ด์–ธํ‹ฐํ”ฝ์ฝ”๋ฆฌ์•„ ์ฃผ์‹ํšŒ์‚ฌ)

           

          ์จ๋ชจ ํ”ผ์…” ์‚ฌ์ด์–ธํ‹ฐํ”ฝ ์†”๋ฃจ์…˜์Šค ์œ ํ•œํšŒ์‚ฌ
          ๋Œ€ํ‘œ์ž : ์„์ˆ˜์ง„
          ์‚ฌ์—…์ž ๋“ฑ๋ก๋ฒˆํ˜ธ : 114-86-04783
          ์ž…๊ธˆ๊ณ„์ขŒ : ์‹ ํ•œ์€ํ–‰ 140-004-396660
          (์˜ˆ๊ธˆ์ฃผ : ์จ๋ชจํ”ผ์…”์‚ฌ์ด์–ธํ‹ฐํ”ฝ์†”๋ฃจ์…˜์Šค ์œ ํ•œํšŒ์‚ฌ)

           

          ์ฃผ์†Œ : ์„œ์šธ์‹œ ๊ฐ•๋‚จ๊ตฌ ๊ด‘ํ‰๋กœ 281 ์ˆ˜์„œ์˜คํ”ผ์Šค๋นŒ๋”ฉ 12์ธต 06349 | ํ†ต์‹ ํŒ๋งค์—…์‹ ๊ณ ๋ฒˆํ˜ธ : 2015-์„œ์šธ๊ฐ•๋‚จ-00898

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-d8hkz:80/100.66.77.44:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0