Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SSVQCKLRYG MWHFLLGDKA SKRLTERSRV ITVDGNICTG KGKLAKEIAE KLGFKHFPEA GIHYPDSTTG DGKPLATDYN GNCSLEKFYD DPRSNDGNSY RLQSWLYSSR LLQYSDALEH LLT
Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 74%.
The protein encoded by this gene is a component of 42 kDa complex I, the first enzyme complex in the electron transport chain of mitochondria. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. A mutation in this gene was found in an individual with Leigh syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: complex I 42kDa subunit; complex I subunit; FLJ21923; MGC5103; NADH-ubiquinone oxidoreductase 42 kDa subunit; NADHDH; unnamed protein product
Gene Aliases: CI-42k; CI-42KD; MC1DN22
UniProt ID: (Human) O95299
Entrez Gene ID: (Human) 4705
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support