Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: PDFRVLNDGS VYTARAVALS DKKRSFTIWL SDKRKQTQKE VTVLLEHQKK VSKTRHTRET
Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 73%.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARVC10; ARVD10; cadherin family member 3; CDHF5; CDHF6; CMD1BB; desmocollin-4; desmoglein-2; desmoglein-3; desmosomal cadherin; HDGC; PVA; unnamed protein product
Gene Aliases: CDHF3; DSC; DSC1; DSC2; DSC4; HT-CP
UniProt ID: (Human) Q14574
Entrez Gene ID: (Human) 1825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support