Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total BEX1/2.
BEX1/BEX2 belongs to the brain expressed X-linked gene family. The encoded protein interacts with the transcription factor LIM domain only 2 in a DNA-binding complex that recognizes the E-box element and promotes transcription. BEX1/BEX2 has been found to be a tumor suppressor that is silenced in human glioma. In breast cancer cells, this gene product modulates apoptosis in response to estrogen and tamoxifen, and enhances the anti-proliferative effect of tamoxifen. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain expressed gene 1; Brain-expressed X-linked protein 1; Brain-expressed X-linked protein 1 homolog; Brain-expressed X-linked protein 2; Brain-expressed X-linked protein 2 homolog; EG2RVC; expression gene 2 in rat visual cortex; hBex2; HGR74-h protein; ovarian granulosa cell 13.0 kDa protein hGR74; Protein BEX1; Protein BEX2; protein hBex2; reduced expression 3; Reduced expression protein 3; REX-3; unnamed protein product
Gene Aliases: AA407839; AL024066; AU018692; BEX1; BEX2; Bex4; DJ79P11.1; EG2RVC; HBEX2; HGR74-h; Rex3
UniProt ID: (Human) Q9HBH7, (Human) Q9BXY8, (Mouse) Q9R224, (Rat) Q3MKQ2, (Rat) Q3MKQ1, (Mouse) Q9WTZ8
Entrez Gene ID: (Human) 55859, (Human) 84707, (Mouse) 19716, (Rat) 501625, (Rat) 363498, (Mouse) 12069
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support