Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MARK Pan Antibodies

          Invitrogen

          Phospho-MARK Pan (Thr215) Polyclonal Antibody

          View all (3) MARK Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-MARK Pan (Thr215) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          Phospho-MARK Pan (Thr215) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-MARK Pan (Thr215) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-MARK Pan (Thr215) Antibody (PA5-105814) in IHC (P)

          Immunohistochemistry analysis of paraffin-embedded Phospho-MARK Pan (Thr215) in human kidney tissue sections. Antigen retrieval was performed using citrate buffer. Samples were blocked with blocking buffer (1.5 hr, 22°C), incubated with Phospho-MARK Pan (Thr215) polyclonal antibody (Product # PA5-105814) using a dilution of 1:200 (1.5 hr, 22°C), followed by HRP conjugated goat anti-rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-MARK Pan (Thr215) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-MARK Pan (Thr215) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-MARK Pan (Thr215) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-MARK Pan (Thr215) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-MARK Pan (Thr215) Polyclonal Antibody

          Product Details

          PA5-105814

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human MARK1(Accession Q9P0L2), corresponding to amino acid residues around phosphorylated Thr215.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2817213

          Product Specific Information

          Antibody detects endogenous levels of MARK1/2/3/4 only when phosphorylated at Thr215.

          Target Information

          MARK1 (MAP/microtubule affinity-regulating kinase 1) belongs to the MARK family of serine/threonine kinases. MARK family protein kinases phosphorylate microtubule-associated proteins and may play a role in cytoskeletal stability. MARK2 refers to MAP/microtubule affinity-regulating kinase 2 isoform a [Homo sapiens]. EMK (ELKL Motif Kinase) is a small family of ser/thr protein kinases involved in the control of cell polarity, microtubule stability and cancer. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1 called MARK2. MARK3 was originally identified as a marker that was induced by treatment with DNA damaging agents, and loss of MARK3 was found with carcinogenesis in the pancreas. MARK4 contains an N terminal serine/threonine kinase domain, a central ubiquitin associated domain, and a C terminal KA1 associated kinase domain. RT PCR analysis detects upregulated expression of the gene for MARK4 in nearly all clinical hepatocellular carcinoma cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: C-TAK1; cdc25C-associated protein kinase 1; ELKL motif kinase 1; ELKL motif kinase 2; ELKL motif serine-threonine protein kinase 3; EMK-1; EMK-2; EMK1; EMK2/MARK3 serine-threonine protein kinase; epididymis secretory sperm binding protein; MAP/microtubule affinity-regulating kinase 1; MAP/microtubule affinity-regulating kinase 2; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MARK4 serine/threonine protein kinase; MPK-10; PAR1 homolog b; PAR1 homolog c; protein kinase; protein kinase STK10; protein serine/threonine kinase; MARK4L; ser/Thr protein kinase PAR-1; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; serine/threonine-protein kinase p78; testicular tissue protein Li 117; unnamed protein product

          View more View less

          Gene Aliases: 1600015G02Rik; 2410090P21Rik; A430080F22Rik; B930025N23Rik; C-TAK1; CTAK1; Emk; EMK-1; EMK1; Emk2; Emk3; ETK-1; ETK1; KP78; MARK; MARK4L; MARK4S; MARKL1; MARKL1L; mKIAA1477; mPar-1c; MPK10; MRD76; PAR-1; Par-1a; Par-1b; Par-1c; PAR-1D; PAR1A; Par1b; Par1c; VIPB

          View more View less

          UniProt ID: (Human) Q9P0L2, (Human) Q7KZI7, (Human) P27448, (Human) Q96L34, (Mouse) Q8VHJ5, (Rat) O08678, (Mouse) Q05512, (Rat) O08679, (Mouse) Q03141, (Rat) Q8VHF0, (Mouse) Q8CIP4

          View more View less

          Entrez Gene ID: (Human) 4139, (Human) 2011, (Human) 4140, (Human) 57787, (Mouse) 226778, (Rat) 117016, (Mouse) 13728, (Rat) 60328, (Mouse) 17169, (Rat) 170577, (Rat) 680407, (Mouse) 232944

          View more View less

          Function(s)
          magnesium ion binding phosphatidylserine binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding phosphatidylinositol-4,5-bisphosphate binding tau protein binding tau-protein kinase activity phosphatidic acid binding protein serine kinase activity RNA binding protein kinase activator activity cadherin binding protein phosphatase binding microtubule binding cytoskeletal adaptor activity gamma-tubulin binding ubiquitin binding nucleotide binding
          Process(es)
          microtubule cytoskeleton organization neuron migration protein phosphorylation cytoskeleton organization positive regulation of gene expression negative regulation of gene expression negative regulation of epithelial to mesenchymal transition regulation of neuron projection development intracellular signal transduction regulation of dendrite development establishment of mitochondrion localization regulation of postsynapse assembly regulation of G2/M transition of mitotic cell cycle negative regulation of hippo signaling negative regulation of protein localization to nucleus microtubule bundle formation nervous system development cilium organization positive regulation of cilium assembly positive regulation of cell cycle regulation of centrosome cycle positive regulation of NLRP3 inflammasome complex assembly positive regulation of protein localization to centrosome mitophagy positive regulation of neuron projection development establishment of cell polarity establishment or maintenance of epithelial cell apical/basal polarity protein autophosphorylation regulation of axonogenesis regulation of cytoskeleton organization mitochondrion localization axon development regulation of microtubule cytoskeleton organization establishment or maintenance of cell polarity regulating cell shape regulation of neurofibrillary tangle assembly positive regulation of programmed cell death regulation of postsynapse organization microtubule polymerization cilium disassembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0