Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Antibody detects endogenous levels of total DST.
As basal cells of stratified squamous epithelia begin to migrate in response to wound healing, they lose their cell-substrate adhesion junctions, the hemidesmosomes. The hemidesmosome is an adhesion structure of the epidermal-dermal junction in keratinocytes. When keratinocytes migrate laterally or upward to differentiate they must control the formation and disintegration of the hemidesmosomes. The bullous pemphigoid antigen BPAG1 is a hemidesmosomal protein of the cutaneous basement membrane zone. The primary sequence deduced from full-length human cDNAs predicts that this molecule consists of a central rod region and flanking globular domains. A neuronal isoform, BPAG1n3 is the result of differential splicing of BPAG1. BPAG1n3 is distinguished by its initial 32 amino acid residues and by the absence of the amino-terminal half of the actin-binding domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPA; BPAG1a3; BPAG1eA; MACF-like product; BPAG1eB; second IF-binding domain product; bullous pemphigoid antigen; bullous pemphigoid antigen 1; cytoskeletal protein; cytoskeletal protein BPAG1; dystonia musculorum protein; hemidesmosomal plaque protein; microtubule actin cross-linking factor 2; trabeculin-beta; unnamed protein product
Gene Aliases: 2310001O04Rik; A830042E19Rik; ah; athetoid; BP230; BP240; BPA; Bpag; BPAG1; BPAG1-n; CATX-15; CATX15; CMYO29; D6S1101; DMH; DT; EBS3; EBSB2; HSAN6; LCCS12; MACF2; mKIAA0728; nmf203; nmf339
UniProt ID: (Human) Q03001, (Mouse) Q91ZU6
Entrez Gene ID: (Human) 667, (Mouse) 13518, (Rat) 316313
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support