Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Antibody detects endogenous levels of total HOP.
HOPX is an atypical homeodomain protein which does not bind DNA and is required to modulate cardiac growth and development. HOPX acts via its interaction with SRF, thereby modulating the expression of SRF-dependent cardiac-specific genes and cardiac development. HOPX prevents SRF-dependent transcription either by inhibiting SRF binding to DNA or by recruiting histone deacetylase (HDAC) proteins that prevent transcription by SRF. HOPX overexpression causes cardiac hypertrophy and may act as a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeobox domain; homeobox only domain; homeobox-only protein; Hop; lung cancer-associated Y protein; mOB1; not expressed in choriocarcinoma clone 1; odd homeobox 1; odd homeobox protein 1; unnamed protein product
Gene Aliases: 1110018K11Rik; 1200015P04Rik; 2300002F06Rik; CAMEO; Hdop; HOD; HOP; LAGY; NECC1; OB1; Obl; SMAP31; TOTO
UniProt ID: (Human) Q9BPY8, (Mouse) Q8R1H0
Entrez Gene ID: (Human) 84525, (Mouse) 74318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support