Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Myosin Heavy Chain Antibodies

          Invitrogen

          Myosin Heavy Chain Polyclonal Antibody

          View all (4) Myosin Heavy Chain antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Myosin Heavy Chain Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          Myosin Heavy Chain Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Myosin Heavy Chain Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Myosin Heavy Chain Antibody (PA5-115216) in ICC/IF

          Immunocytochemistry analysis of Myosin Heavy Chain in Hela cells. The samples were fixed with PFA and permeabilized in 0.1% Triton X-100, then blocked in 10% serum for 45 minutes at 25°C. Samples were incubated with Myosin Heavy Chain polyclonal antibody (Product # PA5-115216) and mouse anti-beta tubulin antibody for 1 hour at 37°C. An AlexaFluor594 conjugated goat anti-rabbit IgG (Red) and an AlexaFluor488... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Myosin Heavy Chain Antibody in Immunocytochemistry (ICC/IF)
          Myosin Heavy Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Myosin Heavy Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Myosin Heavy Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Myosin Heavy Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Myosin Heavy Chain Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Myosin Heavy Chain Polyclonal Antibody

          Product Details

          PA5-115216

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human MYH1(Accession P12882), corresponding to amino acid residues V608-N658.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2899852

          Product Specific Information

          Antibody detects endogenous levels of total Fast Myosin Skeletal Heavy chain.

          Target Information

          The function of Mhc is muscle contraction.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha cardiac MHC; alpha myosin; alpha-myosin heavy chain; beta-myosin heavy chain; cardiac myosin heavy chain 21/26; cardiomyopathy, hypertrophic 1; epididymis luminal protein 71; MHC; molecular motor; muscle myofibrillar protein; myHC-2x; myHC-alpha; myHC-IIx/d; myosin heavy chain; myosin heavy chain 1; myosin heavy chain 2X; myosin heavy chain 6; Myosin heavy chain 7; myosin heavy chain IIx/d; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, skeletal muscle, adult 1; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); type 2X myosin heavy chain; unnamed protein product

          View more View less

          Gene Aliases: A530084A17Rik; A830009F23Rik; alpha-MHC; alphaMHC; ASD3; CMD1EE; CMH14; HEL71; IId; IId/x; MdMs; MHC-2X/D; MHC2X/D; Myh7; MYHa; MYHC; MyHC-2x; MyHC-2X/D; Myhc-a; MyHC-beta; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MyHC-slow; MYHCA; Myhs-f; Myhs-f2; MYHSA1; Myhsf2; Myosin-7; SSS3

          View more View less

          UniProt ID: (Human) P12882, (Human) P13533, (Mouse) Q5SX40, (Mouse) Q02566

          View more View less

          Entrez Gene ID: (Human) 4619, (Human) 4624, (Mouse) 17879, (Rat) 287408, (Mouse) 17888, (Rat) 29556

          View more View less

          Function(s)
          microfilament motor activity motor activity protein binding ATP binding actin filament binding myosin phosphatase activity protein kinase binding calcium-dependent ATPase activity identical protein binding macromolecular complex binding ATPase activity
          Process(es)
          muscle contraction sensory perception of sound in utero embryonic development regulation of the force of heart contraction regulation of heart rate striated muscle contraction adult heart development visceral muscle development regulation of heart contraction regulation of blood pressure cardiac muscle hypertrophy in response to stress actin filament-based movement muscle filament sliding myofibril assembly regulation of ATPase activity sarcomere organization ATP metabolic process atrial cardiac muscle tissue morphogenesis ventricular cardiac muscle tissue morphogenesis cardiac muscle cell development cardiac muscle contraction regulation of heart growth muscle system process cellular response to hydrogen peroxide cellular response to angiotensin cellular response to 3,3',5-triiodo-L-thyronine
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-m8c75:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0