Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive Samples: Human serum
Immunogen sequence: MYGKIIFVLL LSEIVSISAL STTEVAMHTS TSSSVTKSYI SSQTNGETGQ LVHRFTVPAP VVIILIILCV MAGIIGTILL ISYTIRRLIK A
This protein is a minor sialoglycoprotein in erythrocyte membranes.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: blood group system MNSs, Mur-like; blood group system MNSs, St(a) type E; blood group system MNSs, St(a) type F; glycophorin B blood group antigen; glycophorin B precursor; glycophorin B, blood group system Ss, S, Mit+; glycophorin Hop; GPB.NY; GYPHe.NY; hybrid glycophorin Kip; sialoglycoprotein delta; Ss blood group; Ss sialoglycoprotein; SS-active sialoglycoprotein
Gene Aliases: CD235b; GPB; GYP; MNS; PAS-3; SS
UniProt ID: (Human) P06028
Entrez Gene ID: (Human) 2994
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support