Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MYH2 (Myosin-2) is associated with muscle contraction and is required for cytoskeleton organization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2A myosin heavy chain; fast myosin heavy chain; muscle myofibrillar protein; myHC-2a; myosin heavy chain 2; myosin heavy chain 2A; myosin heavy chain IIa; myosin heavy chain type IIx; myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; skeletal muscle myosin heavy chain II-a
Gene Aliases: MHC2A; Myh1; Myh2a; MyHC-2A; MyHC-IIA; Myhs-f; Myhs-f1; Myhsf1
UniProt ID: (Pig) Q9TV63, (Mouse) G3UW82
Entrez Gene ID: (Pig) 397256, (Rat) 691644, (Mouse) 17882
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support