Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 9
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive Control: Mouse heart tissue, rat heart tissue, 293T, HepG2, LOVO, human lung cancer tissue, human pancreas tissue, mouse pancreas tissue.
Subcellular Location: Membrane.
G protein-coupled inwardly rectifying potassium channels (Kir3.1 through Kir3.4) are coupled to numerous neurotransmitter receptors in the brain and are abundantly expressed in the olfactory bulb, hippocampus, neocortex, dentate gyrus, cerebellar cortex and thalamus regions of the brain. Also known as GIRK, Kir3 potassium channels localize to the soma and dendrites as well as axons of neurons. Liberated Gbg subunits from G protein heterotrimers bind to and regulate Kir3 channel activity. Gb3- and Gb4-containing Gbg dimers bind directly to cytoplasmic domains of Kir3 proteins and increase the K+ current while Gb5-containing Gbg dimers inhibit Kir3 K+ current. Kir3 activity is also inhibited by tyrosine phosphorylation. Brain-derived neurotrophic factor activates receptor tyrosine kinase B, which then phosphorylates Kir3 tyrosine residues, effectively inactivating the Kir3 channels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac ATP-sensitive potassium channel; cardiac inward rectifier; CIR; GIRK-4; GIRK4; heart KATP channel; integral membrane protein; inward rectifier K(+) channel Kir3.4; inward rectifier K+ channel KIR3.4; IRK-4; IRK4; KATP-1; potassium channel, inwardly rectifying subfamily J member 5; potassium channel, inwardly rectifying subfamily J, member 5; potassium voltage-gated channel subfamily J member 5; unnamed protein product
Gene Aliases: CIR; GIRK4; KATP-1; KATP1; KIR3.4; LQT13
UniProt ID: (Human) P48544, (Mouse) P48545, (Rat) P48548
Entrez Gene ID: (Human) 3762, (Mouse) 16521, (Rat) 29713
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support