Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive test controls include: mouse brain, mouse testis, rat brain, rat testis. The target is usually found in the following locations: Nucleus.
Immunogen sequence: MEPYSLGEEG ALPSEGHLPS FSESQVLNCS DTLNRDLGPS TRDLLYAGLS GLDLDPSLST SDMPSEVLED NLDTLSLYSG KDSDSVKLLE EYADSESQTS LQDLGLGALK VPKEADEGGR ATGSTRKGKR QHSSPQNPLL DCSLCGKVFS SASSLSKHYL THSQERKHVC KVCSKAFKRQ DHLTGHMLTH QKTKPFVCIE QGCSKSYCDY RSLRRHYEVQ HGVCILKETP PEEEAYGDPT HNHDVANQPP PSGLRSLGPP EARSPGSVLP NRDLLRCIVS SIVHQKIPSP GPAVGPSDTE
Component of some chromatin remodeling multiprotein complex that plays a role during spermatogenesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: spermatogenic cell HDAC-interacting protein 1
Gene Aliases: EG666528; Znf541
UniProt ID: (Mouse) Q0GGX2
Entrez Gene ID: (Mouse) 666528, (Rat) 308108
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support