Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Laminin is a crucial glycoprotein that plays a significant role in the extracellular matrix, particularly within the basement membrane. It is composed of three subunits: alpha, beta, and gamma chains. These chains are disulfide-linked polypeptides that together form a heterotrimer, resembling the shape of a cross. The human genome encodes 11 genetically distinct laminin chains, which combine to form over 14 different laminin isoforms, each with specific tissue distributions and functions. Laminins can bind to other laminin molecules, as well as integrins and other matrix proteins, providing structural support and influencing cell differentiation, migration, and adhesion. These glycoproteins are vital during early embryonic development, where they contribute to the formation of basement membrane-like structures, even in the absence of type IV collagen. The evolutionary origin of laminin gene families can be traced back to early branching animals, providing insights into metazoan gene family development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dystrophia muscularis; laminin A chain; laminin A-chain; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B2 chain; laminin B2 precursor; laminin M chain; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; laminin-1 subunit beta; laminin-1 subunit gamma; laminin-10 subunit beta; laminin-10 subunit gamma; laminin-11 subunit gamma; laminin-111; laminin-12 subunit alpha; laminin-12 subunit beta; laminin-2 subunit alpha; laminin-2 subunit beta; laminin-2 subunit gamma; laminin-3 subunit alpha; laminin-3 subunit gamma; laminin-4 subunit alpha; laminin-4 subunit gamma; laminin-6 subunit beta; laminin-6 subunit gamma; laminin-7 subunit gamma; laminin-8 subunit beta; laminin-8 subunit gamma; laminin-9 subunit gamma; merosin heavy chain; mutant laminin subunit alpha 2; S-LAM alpha; S-LAM gamma; S-laminin subunit alpha; S-laminin subunit gamma; unnamed protein product
Gene Aliases: 5830440B04; B2e; CLM; D130003D08Rik; dy; LAMA; Lamb-1; Lamb1-1; LAMB2; LAMM; LIS5; LKBMH; LUCAO; MDC1A; mer; merosin; mKIAA4087; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391, (Human) P24043, (Human) P07942, (Human) P11047, (Mouse) P19137, (Mouse) Q60675, (Mouse) P02469
Entrez Gene ID: (Human) 284217, (Human) 3908, (Human) 3912, (Human) 3915, (Rat) 316758, (Mouse) 16772, (Rat) 309368, (Mouse) 16773, (Mouse) 16777, (Rat) 298941, (Mouse) 226519, (Rat) 117036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support