Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAF:lysophospholipid transacetylase; PAF:sphingosine transacetylase; platelet-activating factor acetylhydrolase 2, 40kDa; Platelet-activating factor acetylhydrolase 2, cytoplasmic; Platelet-activating factor acetyltransferase PAFAH2; SD-PLA2; Serine-dependent phospholipase A2; unnamed protein product
Gene Aliases: HSD-PLA2; PAFAH2
UniProt ID: (Human) Q99487
Entrez Gene ID: (Human) 5051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support