Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CDC42 Antibodies

          Invitrogen

          Cdc42 Polyclonal Antibody

          View all (17) CDC42 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Cdc42 Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Cdc42 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Cdc42 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cdc42 Antibody (PA5150246) in WB

          Western blot analysis of Cdc42 in recombinant protein. Samples were incubated with Cdc42 Polyclonal (Product # PA5150246) antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cdc42 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Cdc42 Polyclonal Antibody

          Product Details

          PA5150246

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant Cell Division Cycle Protein 42 (CDC42) (Met1-Cys188)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3716281

          Target Information

          Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for the Rac1 gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cdc42; CDC42Mm; cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; GTP-binding protein; GTP-binding protein G25K; RP1-224A6.5; small GTP binding protein; small GTP binding protein CDC42; small GTP binding protein; GTPase; unnamed protein product

          View more View less

          Gene Aliases: CDC42Hs; G25K; TKS

          View more View less

          UniProt ID: (Human) P60953, (Mouse) P60766, (Rat) Q8CFN2

          View more View less

          Entrez Gene ID: (Human) 998, (Mouse) 12540, (Rat) 64465

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding protein kinase binding GTP-dependent protein binding thioesterase binding GBD domain binding apolipoprotein A-I receptor binding identical protein binding ubiquitin protein ligase activity mitogen-activated protein kinase kinase kinase binding
          Process(es)
          heart process cardiac neural crest cell migration involved in outflow tract morphogenesis endocytosis phagocytosis, engulfment actin filament organization Golgi organization establishment or maintenance of cell polarity signal transduction integrin-mediated signaling pathway small GTPase mediated signal transduction anatomical structure morphogenesis regulation of lamellipodium assembly positive regulation of lamellipodium assembly substantia nigra development establishment of cell polarity actin cytoskeleton organization macrophage differentiation positive regulation of cell growth positive regulation of cell migration positive regulation of pseudopodium assembly negative regulation of protein complex assembly positive regulation of cytokinesis regulation of actin cytoskeleton organization cell junction assembly embryonic heart tube development neuropilin signaling pathway modulation by host of viral process establishment of epithelial cell apical/basal polarity positive regulation of pinocytosis positive regulation of cellular component organization regulation of filopodium assembly positive regulation of filopodium assembly regulation of stress fiber assembly positive regulation of stress fiber assembly establishment of Golgi localization regulation of attachment of spindle microtubules to kinetochore Wnt signaling pathway, planar cell polarity pathway positive regulation of epithelial cell proliferation involved in lung morphogenesis dendritic spine morphogenesis organelle transport along microtubule endothelin receptor signaling pathway regulation of postsynapse organization positive regulation of substrate adhesion-dependent cell spreading sprouting angiogenesis cardiac conduction system development protein phosphorylation regulation of mitotic nuclear division nuclear migration Rho protein signal transduction protein localization endosomal transport adherens junction organization establishment or maintenance of apical/basal cell polarity dendritic cell migration positive regulation of catalytic activity positive regulation of MAPK cascade positive regulation of neuron apoptotic process positive regulation of DNA replication positive regulation of JNK cascade filopodium assembly neuron fate determination nucleus localization positive regulation of synapse structural plasticity positive regulation of protein kinase B signaling heart contraction cellular response to interferon-gamma actin filament branching positive regulation of intracellular protein transport cell-cell adhesion regulation of modification of postsynaptic structure response to glucose regulation of exocytosis submandibular salivary gland formation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-gh2m7:80/100.66.74.151:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0