Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
It is not recommended to aliquot this antibody.
This gene encodes a protein that interacts with the DNA polymerase delta p50 subunit. This protein is a specific target of S6 kinase 1 and regulates cell growth. Two transcript variants that encode different protein isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 46 kDa DNA polymerase delta interaction protein; dJ222E13.3; polymerase (DNA) delta interacting protein 3; polymerase (DNA-directed), delta interacting protein 3; polymerase delta interacting protein 46; RNA-binding protein P46; S6K1 Aly/REF-like target; unnamed protein product
Gene Aliases: 1110008P04Rik; mKIAA1649; PDIP3; PDIP46; SKAR
UniProt ID: (Human) Q9BY77, (Mouse) Q8BG81
Entrez Gene ID: (Human) 84271, (Rat) 315170, (Mouse) 73826
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support