Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 10
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody does not react with MARK4.
It is not recommended to aliquot this antibody.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-TAK1; cdc25C-associated protein kinase 1; ELKL motif kinase 1; ELKL motif kinase 2; ELKL motif serine-threonine protein kinase 3; EMK-1; EMK-2; EMK1; EMK2/MARK3 serine-threonine protein kinase; MAP/microtubule affinity-regulating kinase 1; MAP/microtubule affinity-regulating kinase 2; MPK-10; PAR1 homolog b; PAR1 homolog c; protein kinase; protein kinase STK10; ser/Thr protein kinase PAR-1; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; serine/threonine-protein kinase p78; testicular tissue protein Li 117; unnamed protein product
Gene Aliases: 1600015G02Rik; A430080F22Rik; B930025N23Rik; C-TAK1; CTAK1; Emk; EMK-1; EMK1; Emk2; Emk3; ETK-1; ETK1; KP78; MARK; mKIAA1477; mPar-1c; MPK10; MRD76; PAR-1; Par-1a; Par-1b; Par-1c; PAR1A; Par1b; Par1c; VIPB
UniProt ID: (Human) Q9P0L2, (Human) Q7KZI7, (Human) P27448, (Mouse) Q8VHJ5, (Rat) O08678, (Mouse) Q05512, (Rat) O08679, (Mouse) Q03141, (Rat) Q8VHF0
Entrez Gene ID: (Human) 4139, (Human) 2011, (Human) 4140, (Mouse) 226778, (Rat) 117016, (Mouse) 13728, (Rat) 60328, (Mouse) 17169, (Rat) 170577
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support