Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • UBE2A/UBE2B Antibodies

          Invitrogen

          UBE2A/UBE2B Polyclonal Antibody

          View all (2) UBE2A/UBE2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UBE2A/UBE2B Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          UBE2A/UBE2B Polyclonal Antibody

          Product Details

          PA5-17592

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Flow Cytometry (Flow)

          1:50
          -
          Product Specifications

          Species Reactivity

          Chicken, Fruit fly, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to amino acids at the amino-terminus of human HR6A
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          200 µg/mL

          Amount

          20 µg

          Purification

          Antigen affinity chromatography, Protein A

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10981771

          Product Specific Information

          It is not recommended to aliquot this antibody.

          Target Information

          The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E2 protein; E2 ubiquitin-conjugating enzyme A; E2 ubiquitin-conjugating enzyme B; E2(14k); HHR6A (Human homologue of yeast RAD 6); putative; HHR6B (Human homologue of yeast RAD 6); putative; RAD6 homolog A; RAD6 homolog B; UBE2B; ubiquitin carrier protein A; ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2B; ubiquitin-conjugating enzyme E2(14k); ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2A; ubiquitin-conjugating enzyme E2A (RAD6 homolog); ubiquitin-conjugating enzyme E2B; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-conjugating enzyme HR6A; ubiquitin-protein ligase A; ubiquitin-protein ligase B; unnamed protein product

          View more View less

          Gene Aliases: E2-14k; E2-17kDa; E214K; HHR6A; HHR6B; HR6A; HR6B; Mhr6a; mHR6B; mp:zf637-2-000771; MRXS30; MRXSN; RAD6A; RAD6B; si:bz46j2.4; UBC2; wu:fa01h11; zgc:100921

          View more View less

          UniProt ID: (Human) P49459, (Human) P63146, (Mouse) Q9Z255, (Mouse) P63147, (Rat) P63149

          View more View less

          Entrez Gene ID: (Chicken) 395672, (Human) 7319, (Chicken) 100857678, (Human) 7320, (Rat) 298317, (Mouse) 22209, (Mouse) 22210, (Rat) 81816, (Zebrafish) 797853, (Zebrafish) 437020

          View more View less

          Function(s)
          ubiquitin-protein transferase activity protein binding ubiquitin-like protein transferase activity ubiquitin protein ligase binding ubiquitin binding ubiquitin conjugating enzyme activity ATP binding nucleotide binding transferase activity
          Process(es)
          G2/M transition of mitotic cell cycle protein polyubiquitination in utero embryonic development blastocyst hatching DNA repair chromatin remodeling biological_process response to UV protein ubiquitination proteasome-mediated ubiquitin-dependent protein catabolic process maternal process involved in female pregnancy protein K48-linked ubiquitination protein K11-linked ubiquitination positive regulation of mitophagy postreplication repair chromatin organization ubiquitin-dependent protein catabolic process cellular response to DNA damage stimulus spermatogenesis sperm axoneme assembly response to xenobiotic stimulus positive regulation of reciprocal meiotic recombination negative regulation of apoptotic process meiotic telomere clustering protein stabilization chiasma assembly protein autoubiquitination synaptonemal complex organization protein K63-linked ubiquitination positive regulation of canonical Wnt signaling pathway negative regulation of post-translational protein modification response to hypoxia cellular response to insulin stimulus histone ubiquitination
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-x4rxd:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0