Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
It is not recommended to aliquot this antibody.
This antibody is not cross-reactive with endogenous levels of other PKC isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atypical protein kinase C; nPKC-eta; nPKC-zeta; PKC-L; PRKCI; protein kinase C zeta subspecies; protein kinase C, zeta subspecies; Protein uPEP2; unnamed protein product
Gene Aliases: 14-3-3-zetaisoform; aPKCzeta; nPKC-eta; nPKC-zeta; PKC-L; PKC-ZETA; PKC2; Pkch; PKCL; Pkcz; PRKCL; r14-3-3; uORF2; zetaPKC
UniProt ID: (Human) P24723, (Human) Q05513, (Mouse) P23298, (Rat) Q64617, (Mouse) Q02956, (Rat) P09217
Entrez Gene ID: (Human) 5583, (Human) 5590, (Mouse) 18755, (Rat) 81749, (Mouse) 18762, (Rat) 25522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support