Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
A suggested positive control is human brain tissue lysate.
PA5-20520 can be used with blocking peptide PEP-0640.
Lipogenesis is a key event in the energy storage system and is controlled by the transcription factor sterol regulatory element-binding protein (SREBP)-1. Elongation of very long chain fatty acids protein 6 (ELOVL6) is a member of fatty acyl-CoA elongase gene family that converts palmitic to stearic acid and it has been shown to be a target of SREBP-1, playing an important role in de novo synthesis of long-chain saturated and monosaturated fatty acids in conjunction with fatty acid synthase and stearoyl-CoA desaturase ELOVL6 was predicted to be important for tissue fatty acid composition. Recent studies suggest that inhibition of this elongase could be a new therapeutic approach for ameliorating insulin resistance, diabetes and cardiovascular risks, even in the presence of a continuing state of obesity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-keto acyl-CoA synthase ELOVL6; elongation of very long chain fatty acids protein 6; elongation of very long chain fatty acids protein 6-like; ELOVL FA elongase 6; ELOVL family member 6, elongation of long chain fatty acids; ELOVL family member 6, elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast); fatty acid elongase 2; fatty acyl-CoA elongase; long chain fatty acid elongase; long chain fatty acid elongation enzyme; long chain fatty acyl elongase; long-chain fatty-acyl elongase; myelin-associated SUR4 protein; unnamed protein product; very long chain 3-ketoacyl-CoA synthase 6; very long chain 3-oxoacyl-CoA synthase 6
Gene Aliases: FACE; FAE; hELO2; LCE; Lce2; rELO2
UniProt ID: (Human) Q9H5J4, (Mouse) Q920L5, (Rat) Q920L6
Entrez Gene ID: (Human) 79071, (Mouse) 170439, (Rat) 171402
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support